Product Details

SNP ID
rs660392
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:72329662 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGTCCTGAGGCCCAAACCCTGCCT[C/G]TCAGCTGCCTCCTGCCCTACAAACT
Phenotype
MIM: 300269
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HDAC8 PubMed Links

Gene Details

Gene
HDAC8
Gene Name
histone deacetylase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166418.1 1868 UTR 3 NP_001159890.1
NM_001166419.1 1868 Intron NP_001159891.1
NM_001166420.1 1868 Intron NP_001159892.1
NM_001166422.1 1868 Intron NP_001159894.1
NM_001166448.1 1868 Intron NP_001159920.1
NM_018486.2 1868 UTR 3 NP_060956.1
XM_011530986.2 1868 UTR 3 XP_011529288.2
XM_011530987.2 1868 Intron XP_011529289.2
XM_017029640.1 1868 UTR 3 XP_016885129.1
XM_017029641.1 1868 UTR 3 XP_016885130.1
XM_017029642.1 1868 UTR 3 XP_016885131.1
XM_017029643.1 1868 UTR 3 XP_016885132.1
XM_017029644.1 1868 UTR 3 XP_016885133.1
XM_017029645.1 1868 UTR 3 XP_016885134.1
XM_017029646.1 1868 UTR 3 XP_016885135.1
XM_017029647.1 1868 Intron XP_016885136.1

View Full Product Details