Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005967.3 | 3179 | Intron | NP_005958.1 | ||
XM_005268894.2 | 3179 | Intron | XP_005268951.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001178078.1 | 3179 | UTR 3 | NP_001171549.1 | ||
NM_001178079.1 | 3179 | UTR 3 | NP_001171550.1 | ||
NM_001178080.1 | 3179 | UTR 3 | NP_001171551.1 | ||
NM_001178081.1 | 3179 | UTR 3 | NP_001171552.1 | ||
NM_003153.4 | 3179 | UTR 3 | NP_003144.3 | ||
XM_006719575.2 | 3179 | UTR 3 | XP_006719638.1 | ||
XM_011538703.2 | 3179 | UTR 3 | XP_011537005.1 | ||
XM_011538704.2 | 3179 | UTR 3 | XP_011537006.1 | ||
XM_011538705.2 | 3179 | UTR 3 | XP_011537007.1 | ||
XM_011538706.2 | 3179 | UTR 3 | XP_011537008.1 | ||
XM_011538707.2 | 3179 | UTR 3 | XP_011537009.1 | ||
XM_011538708.2 | 3179 | UTR 3 | XP_011537010.1 | ||
XM_011538709.2 | 3179 | UTR 3 | XP_011537011.1 |