Product Details
- SNP ID
-
rs630865
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:31373589 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTATTTCGGTTATTCTAGTTTGCTT[A/G]TATTTCCACAGAAATTTTAGAATCA
- Phenotype
-
MIM: 608062
MIM: 611072
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DCDC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76441383] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DCDC1
- Gene Name
- doublecortin domain containing 1
There are no transcripts associated with this gene.
- Gene
- DNAJC24
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C24
View Full Product Details