Product Details

SNP ID
rs487178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31370402 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCTTACTTGTGTGCCAGTTTTAT[C/T]ATAAGACTAGCAGAAATACCTTCTT
Phenotype
MIM: 608062 MIM: 611072
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCDC1 PubMed Links

Gene Details

Gene
DCDC1
Gene Name
doublecortin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181807.3 Intron NP_861523.2
Gene
DNAJC24
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181706.4 Intron NP_859057.4
XM_017017216.1 Intron XP_016872705.1

View Full Product Details