Product Details

SNP ID
hCV621250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110282770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAGACTTGCTAGTTAGGATTT[C/T]ATTACTGGCAGCATGTATATCTTTT
Phenotype
MIM: 108740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP2A2 PubMed Links

Gene Details

Gene
ATP2A2
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001681.3 757 Missense Mutation TCA,TTA S65L NP_001672.1
NM_170665.3 757 Missense Mutation TCA,TTA S65L NP_733765.1

View Full Product Details