Product Details
- SNP ID
-
rs314262
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:104946746 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGCTTTCCAAAGAATGTAGTTCAGT[A/G]ATAGTTGACACATACTATAGATTAA
- Phenotype
-
MIM: 611044
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LIN28B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73771026] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LIN28B
- Gene Name
- lin-28 homolog B
- Gene
- LIN28B-AS1
- Gene Name
- LIN28B antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details