Product Details

SNP ID
rs580933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65429413 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGACGGATCGAGTCTGCTTTCCCT[C/G]TGGATGTGAGCATTGCTTTGTCTGC
Phenotype
MIM: 612769
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NEAT1 PubMed Links

Gene Details

Gene
NEAT1
Gene Name
nuclear paraspeckle assembly transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details