Product Details

SNP ID
rs684217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:2851752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAACCAGGAACAGAAGGGGTTT[C/T]TATATAAGTGGGCATTTTAAGTAAT
Phenotype
MIM: 608928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMILIN2 PubMed Links

Gene Details

Gene
EMILIN2
Gene Name
elastin microfibril interfacer 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032048.2 Intron NP_114437.2
XM_017026038.1 Intron XP_016881527.1

View Full Product Details