Product Details

SNP ID
rs377205432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:128839289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTAAAGTTATGGAAATCCACTC[A/G]GTATTTCCCTTCCTTTGTCTTGGAT
Phenotype
MIM: 600359
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNJ1 PubMed Links

Gene Details

Gene
KCNJ1
Gene Name
potassium voltage-gated channel subfamily J member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000220.4 1347 Nonsense Mutation CGA,TGA R338* NP_000211.1
NM_153764.2 1347 Nonsense Mutation CGA,TGA R319* NP_722448.1
NM_153765.2 1347 Nonsense Mutation CGA,TGA R336* NP_722449.3
NM_153766.2 1347 Nonsense Mutation CGA,TGA R319* NP_722450.1
NM_153767.3 1347 Nonsense Mutation CGA,TGA R319* NP_722451.1

View Full Product Details