Product Details
- SNP ID
-
rs288322
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:182836139 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTGATTCTCAACTTCCCAAGGAAG[A/T]CCAAACATCTTGAATCGTCTGCTAT
- Phenotype
-
MIM: 605083
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FRZB
PubMed Links
Gene Details
- Gene
- FRZB
- Gene Name
- frizzled-related protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001463.3 |
|
Intron |
|
|
NP_001454.2 |
View Full Product Details