Product Details
- SNP ID
-
rs300568
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:123397011 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGACTTTTGCAAGGAACTGAACATT[G/T]ATTTATTTGAACTTGCTTTCCAGCT
- Phenotype
-
MIM: 602465
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SPRY1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74448744] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPRY1
- Gene Name
- sprouty RTK signaling antagonist 1
View Full Product Details