Product Details

SNP ID
rs167723
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:146592626 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTGATCTGGAAAGCCTACTTGGCA[G/T]TAATCTGAACTGTTCAGGCTGGAAT
Phenotype
MIM: 604325
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP2R2B PubMed Links

Gene Details

Gene
PPP2R2B
Gene Name
protein phosphatase 2 regulatory subunit Bbeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271899.1 Intron NP_001258828.1
NM_001271900.1 Intron NP_001258829.1
NM_001271948.1 Intron NP_001258877.1
NM_181674.2 Intron NP_858060.2
NM_181675.3 Intron NP_858061.2
NM_181676.2 Intron NP_858062.1
NM_181677.2 Intron NP_858063.1
NM_181678.2 Intron NP_858064.1

View Full Product Details