Product Details

SNP ID
rs76542238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102839194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCTTTACTTTATTTTCTGGAGG[G/T]CACTGCAAAGGATTCCAATTTCACC
Phenotype
MIM: 612349
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PAH PubMed Links

Gene Details

Gene
PAH
Gene Name
phenylalanine hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000277.1 1812 Missense Mutation GAC,GCC D447A NP_000268.1
XM_017019370.1 1812 Intron XP_016874859.1

View Full Product Details