Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001261406.1 | 6977 | UTR 3 | NP_001248335.1 | ||
NM_001261407.1 | 6977 | UTR 3 | NP_001248336.1 | ||
NM_003829.4 | 6977 | UTR 3 | NP_003820.2 | ||
XM_005251622.4 | 6977 | UTR 3 | XP_005251679.1 | ||
XM_005251623.4 | 6977 | UTR 3 | XP_005251680.1 | ||
XM_006716885.3 | 6977 | UTR 3 | XP_006716948.1 | ||
XM_006716886.3 | 6977 | UTR 3 | XP_006716949.1 | ||
XM_006716887.3 | 6977 | UTR 3 | XP_006716950.1 | ||
XM_006716888.3 | 6977 | UTR 3 | XP_006716951.1 | ||
XM_006716889.3 | 6977 | UTR 3 | XP_006716952.1 | ||
XM_006716891.3 | 6977 | UTR 3 | XP_006716954.1 | ||
XM_017015252.1 | 6977 | UTR 3 | XP_016870741.1 | ||
XM_017015253.1 | 6977 | UTR 3 | XP_016870742.1 | ||
XM_017015254.1 | 6977 | UTR 3 | XP_016870743.1 | ||
XM_017015255.1 | 6977 | UTR 3 | XP_016870744.1 | ||
XM_017015256.1 | 6977 | UTR 3 | XP_016870745.1 | ||
XM_017015257.1 | 6977 | UTR 3 | XP_016870746.1 | ||
XM_017015258.1 | 6977 | Intron | XP_016870747.1 | ||
XM_017015259.1 | 6977 | Intron | XP_016870748.1 |