Product Details

SNP ID
rs591579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:95877962 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACTGGAAGGAAAAGACAAAAAAC[A/C]AGTGGACATGATAATTAAATGTTAT
Phenotype
MIM: 615667
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ERCC6L2 PubMed Links

Gene Details

Gene
ERCC6L2
Gene Name
ERCC excision repair 6 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010895.2 Intron NP_001010895.1
NM_020207.4 Intron NP_064592.2
XM_011518641.2 Intron XP_011516943.1
XM_011518644.2 Intron XP_011516946.1
XM_011518645.2 Intron XP_011516947.1
XM_011518646.2 Intron XP_011516948.1
XM_011518647.1 Intron XP_011516949.1
XM_011518648.2 Intron XP_011516950.1
XM_011518650.2 Intron XP_011516952.1
XM_017014707.1 Intron XP_016870196.1
Gene
LINC00476
Gene Name
long intergenic non-protein coding RNA 476
There are no transcripts associated with this gene.

View Full Product Details