Product Details

SNP ID
rs392818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72577173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGGTCACCCCTGTGCTCTCAGA[A/G]AACAAATTACAAAGTCTCCGTGAGG
Phenotype
MIM: 602658
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01537 PubMed Links

Gene Details

Gene
LINC01537
Gene Name
long intergenic non-protein coding RNA 1537
There are no transcripts associated with this gene.

Gene
PDE2A
Gene Name
phosphodiesterase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143839.3 3516 UTR 3 NP_001137311.1
NM_001146209.2 3516 UTR 3 NP_001139681.1
NM_001243784.1 3516 UTR 3 NP_001230713.1
NM_002599.4 3516 UTR 3 NP_002590.1
XM_005274040.2 3516 UTR 3 XP_005274097.1
XM_011545096.2 3516 UTR 3 XP_011543398.1
XM_017017910.1 3516 UTR 3 XP_016873399.1

View Full Product Details