Product Details

SNP ID
rs430431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:128274141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACCATTTTATACTTTCAAATGTG[A/T]CTTCTATATAAGAATATGGAACACA
Phenotype
MIM: 607735
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PGRMC2 PubMed Links

Gene Details

Gene
PGRMC2
Gene Name
progesterone receptor membrane component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006320.4 Intron NP_006311.2
XM_011531533.1 Intron XP_011529835.1

View Full Product Details