Product Details

SNP ID
rs695061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49868439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAGAAATCTTGCCTCCCAGCTC[C/T]TGTCTGGAAGATAGAAACCTCAGAG
Phenotype
MIM: 604306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAIM2 PubMed Links

Gene Details

Gene
FAIM2
Gene Name
Fas apoptotic inhibitory molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012306.3 3308 UTR 3 NP_036438.2
XM_005268730.3 3308 UTR 3 XP_005268787.1
XM_017019040.1 3308 UTR 3 XP_016874529.1

View Full Product Details