Product Details

SNP ID
rs320152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64000157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGCACTCCTGTGGGGGCCGCA[G/C]CCCTGATGTCCCAGTGACACACGGG
Phenotype
MIM: 610400 MIM: 608337
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
MACROD1 PubMed Links

Gene Details

Gene
MACROD1
Gene Name
MACRO domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014067.3 Intron NP_054786.2
XM_005273939.4 Intron XP_005273996.1
XM_005273940.2 Intron XP_005273997.1
XM_006718521.3 Intron XP_006718584.1
XM_006718522.2 Intron XP_006718585.1
XM_011544970.2 Intron XP_011543272.1
Gene
OTUB1
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 1
There are no transcripts associated with this gene.

View Full Product Details