Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001317199.1 | Intron | NP_001304128.1 | |||
NM_001317200.1 | Intron | NP_001304129.1 | |||
NM_001317201.1 | Intron | NP_001304130.1 | |||
NM_001317202.1 | Intron | NP_001304131.1 | |||
NM_001317203.1 | Intron | NP_001304132.1 | |||
NM_001317204.1 | Intron | NP_001304133.1 | |||
NM_001317205.1 | Intron | NP_001304134.1 | |||
NM_001317206.1 | Intron | NP_001304135.1 | |||
NM_001317207.1 | Intron | NP_001304136.1 | |||
NM_001317208.1 | Intron | NP_001304137.1 | |||
NM_001317209.1 | Intron | NP_001304138.1 | |||
NM_001317210.1 | Intron | NP_001304139.1 | |||
NM_001317211.1 | Intron | NP_001304140.1 | |||
NM_016463.8 | Intron | NP_057547.5 | |||
XM_017009573.1 | Intron | XP_016865062.1 |