Product Details

SNP ID
rs659984
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:38010502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGGAAAGCCAGGGCTGCAGGCA[G/T]TTCCAGCACCAACTCAGACCGCATC
Phenotype
MIM: 194521
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF33A PubMed Links

Gene Details

Gene
ZNF33A
Gene Name
zinc finger protein 33A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278170.1 Intron NP_001265099.1
NM_001278171.1 Intron NP_001265100.1
NM_001278173.1 Intron NP_001265102.1
NM_001278174.1 Intron NP_001265103.1
NM_001278175.1 Intron NP_001265104.1
NM_001278176.1 Intron NP_001265105.1
NM_001278177.1 Intron NP_001265106.1
NM_001278178.1 Intron NP_001265107.1
NM_001278179.1 Intron NP_001265108.1
NM_001324175.1 Intron NP_001311104.1
NM_001324176.1 Intron NP_001311105.1
NM_001324177.1 Intron NP_001311106.1
NM_001324178.1 Intron NP_001311107.1
NM_006954.1 Intron NP_008885.1
NM_006974.2 Intron NP_008905.1
XM_011519650.2 Intron XP_011517952.1
XM_011519651.2 Intron XP_011517953.1
XM_011519653.2 Intron XP_011517955.1
XM_011519654.2 Intron XP_011517956.1
XM_017016617.1 Intron XP_016872106.1

View Full Product Details