Product Details
- SNP ID
-
rs727748
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:86082040 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGATAACAGGAGCACGCTTCCTG[A/T]TTACTCCTCAGTTGATCAGAGTTTG
- Phenotype
-
MIM: 615496
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AGBL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75068562] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGBL1
- Gene Name
- ATP/GTP binding protein like 1
- Gene
- LINC01584
- Gene Name
- long intergenic non-protein coding RNA 1584
There are no transcripts associated with this gene.
View Full Product Details