Product Details

SNP ID
rs209751
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:22567415 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CTACATTGCCTGAGAGGGAGGGAGG[C/G]AGGCGGGAAGCCTTCTCTCTGCCTC
Phenotype
MIM: 176945
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EPHA8 PubMed Links

Gene Details

Gene
EPHA8
Gene Name
EPH receptor A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011540969.1 Intron XP_011539271.1
XM_011540970.1 Intron XP_011539272.1
XM_011540972.1 Intron XP_011539274.1
XM_011540973.1 Intron XP_011539275.1
XM_011540975.2 Intron XP_011539277.1

View Full Product Details