Product Details

SNP ID
rs296567
Assay Type
Validated
NCBI dbSNP Submissions
34
Location
Chr.1:200972806 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
GGCCCCCAAGAGCTGAGGGAAAGAA[C/T]ATAGGGGAAGAAACTTCAAGTCCGT
Phenotype
MIM: 608322
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF21B PubMed Links

Gene Details

Gene
KIF21B
Gene Name
kinesin family member 21B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252100.1 6481 UTR 3 NP_001239029.1
NM_001252102.1 6481 UTR 3 NP_001239031.1
NM_001252103.1 6481 UTR 3 NP_001239032.1
NM_017596.3 6481 UTR 3 NP_060066.2
XM_017000731.1 6481 Intron XP_016856220.1
XM_017000732.1 6481 Intron XP_016856221.1
Gene
MROH3P
Gene Name
maestro heat like repeat family member 3, pseudogene
There are no transcripts associated with this gene.

View Full Product Details