Product Details

SNP ID
rs480008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:46810235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAATTATAAAAGCACAGAGATA[A/T]CAGTTATAAGAACCAAGTAAATAAA
Phenotype
MIM: 603567
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PIAS2 PubMed Links

Gene Details

Gene
PIAS2
Gene Name
protein inhibitor of activated STAT 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324046.1 4237 UTR 3 NP_001310975.1
NM_001324047.1 4237 UTR 3 NP_001310976.1
NM_001324048.1 4237 UTR 3 NP_001310977.1
NM_001324049.1 4237 UTR 3 NP_001310978.1
NM_001324051.1 4237 Intron NP_001310980.1
NM_001324052.1 4237 Intron NP_001310981.1
NM_001324053.1 4237 Intron NP_001310982.1
NM_001324054.1 4237 Intron NP_001310983.1
NM_001324055.1 4237 Intron NP_001310984.1
NM_001324057.1 4237 Intron NP_001310986.1
NM_001324058.1 4237 Intron NP_001310987.1
NM_001324059.1 4237 Intron NP_001310988.1
NM_001324060.1 4237 Intron NP_001310989.1
NM_004671.4 4237 UTR 3 NP_004662.2
NM_173206.3 4237 Intron NP_775298.1
XM_005258377.3 4237 Intron XP_005258434.1
XM_005258379.3 4237 Intron XP_005258436.1
XM_006722571.3 4237 UTR 3 XP_006722634.1
XM_006722572.3 4237 Intron XP_006722635.1
XM_006722573.2 4237 Intron XP_006722636.1
XM_011526258.2 4237 Intron XP_011524560.1
XM_011526259.2 4237 Intron XP_011524561.1
XM_017026068.1 4237 Intron XP_016881557.1
XM_017026069.1 4237 Intron XP_016881558.1
XM_017026070.1 4237 Intron XP_016881559.1
XM_017026071.1 4237 Intron XP_016881560.1
XM_017026072.1 4237 Intron XP_016881561.1

View Full Product Details