Product Details

SNP ID
rs530324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27633669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCCCGGGTTCGAGGCTGGGCT[C/G]TGGGGCTGCGGGGGCGCACGGGCGA
Phenotype
MIM: 602728
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCARA3 PubMed Links

Gene Details

Gene
SCARA3
Gene Name
scavenger receptor class A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016240.2 5 Intron NP_057324.2
NM_182826.1 5 Intron NP_878185.1
XM_017013535.1 5 UTR 5 XP_016869024.1
XM_017013536.1 5 Intron XP_016869025.1
XM_017013537.1 5 Intron XP_016869026.1

View Full Product Details