Product Details

SNP ID
rs685967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30215541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGGATGAGCACCACCATGCCCG[C/T]GATGATGAAGATGATTCCAGCCGTC
Phenotype
MIM: 611231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN8 PubMed Links

Gene Details

Gene
CLDN8
Gene Name
claudin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199328.2 611 Missense Mutation CCG,TCG P129S NP_955360.1
Gene
LINC00307
Gene Name
long intergenic non-protein coding RNA 307
There are no transcripts associated with this gene.

View Full Product Details