Product Details

SNP ID
rs654577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42494925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGTTATTCACTTCTAAATAATG[C/T]ATAATTCCAATTTTTACTTCCTTAC
Phenotype
MIM: 192321
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VIPR1 PubMed Links

Gene Details

Gene
VIPR1
Gene Name
vasoactive intestinal peptide receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251882.1 Intron NP_001238811.1
NM_001251883.1 Intron NP_001238812.1
NM_001251884.1 Intron NP_001238813.1
NM_001251885.1 Intron NP_001238814.1
NM_004624.3 Intron NP_004615.2
XM_005265437.2 Intron XP_005265494.1
XM_005265438.3 Intron XP_005265495.1
XM_005265439.2 Intron XP_005265496.1
XM_011534079.1 Intron XP_011532381.1
XM_011534080.2 Intron XP_011532382.1

View Full Product Details