Product Details

SNP ID
hCV757891
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154779349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTTCTGCAGCTGCTGCAGGGCT[C/T]CTGTCTGCAAAGAAGAAAGCCACAG
Phenotype
MIM: 300126 MIM: 305360
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DKC1 PubMed Links

Gene Details

Gene
DKC1
Gene Name
dyskerin pseudouridine synthase 1
There are no transcripts associated with this gene.

Gene
MPP1
Gene Name
membrane palmitoylated protein 1
There are no transcripts associated with this gene.

Gene
SNORA56
Gene Name
small nucleolar RNA, H/ACA box 56
There are no transcripts associated with this gene.

View Full Product Details