Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001290332.1 | 791 | Intron | NP_001277261.1 | ||
NM_001290333.1 | 791 | Intron | NP_001277262.1 | ||
NM_001290334.1 | 791 | Intron | NP_001277263.1 | ||
NM_138567.4 | 791 | Intron | NP_612634.3 | ||
XM_011520455.1 | 791 | Missense Mutation | ACA,ATA | T142I | XP_011518757.1 |
XM_011520456.2 | 791 | Missense Mutation | ACA,ATA | T142I | XP_011518758.1 |
XM_011520457.2 | 791 | Intron | XP_011518759.1 | ||
XM_017018528.1 | 791 | Missense Mutation | ACA,ATA | T142I | XP_016874017.1 |
XM_017018529.1 | 791 | Intron | XP_016874018.1 |