Product Details
- SNP ID
-
rs583089
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:150974974 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAAACAAGGGGGTGCCAGTGCCCC[C/T]GATAAGTGGTTATTTGGGCATATAT
- Phenotype
-
MIM: 606397
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLRN1
PubMed Links
Gene Details
- Gene
- CLRN1
- Gene Name
- clarin 1
There are no transcripts associated with this gene.
- Gene
- CLRN1-AS1
- Gene Name
- CLRN1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details