Product Details

SNP ID
rs190103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23570934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGACAGAGGATCTCACCGGCTG[A/G]GCAGGCCTCTCGGTTGCAGCGGGCT
Phenotype
MIM: 603534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP1G2 PubMed Links

Gene Details

Gene
AP1G2
Gene Name
adaptor related protein complex 1 gamma 2 subunit
There are no transcripts associated with this gene.

Gene
JPH4
Gene Name
junctophilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146028.1 2715 Silent Mutation NP_001139500.1
NM_032452.2 2715 Silent Mutation NP_115828.2
Gene
LOC102724814
Gene Name
uncharacterized LOC102724814
There are no transcripts associated with this gene.

View Full Product Details