Product Details

SNP ID
rs601372
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101831027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGTTTGTTCAGATATTTAGCA[A/C]CATGCAGGCACTTTTAATATTGGTA
Phenotype
MIM: 608004
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
NFKBIZ PubMed Links

Gene Details

Gene
NFKBIZ
Gene Name
NFKB inhibitor zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005474.2 Intron NP_001005474.1
NM_031419.3 Intron NP_113607.1
Gene
NXPE3
Gene Name
neurexophilin and PC-esterase domain family member 3
There are no transcripts associated with this gene.

View Full Product Details