Product Details
- SNP ID
-
rs480299
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
81
- Location
-
Chr.1:52854602 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGAGTTTCCCTCAGGAAGTAGCCGA[G/T]CGATTTCTCAGGGTGATGACTTGGC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZYG11A
PubMed Links
Gene Details
- Gene
- ZYG11A
- Gene Name
- zyg-11 family member A, cell cycle regulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004339.2 |
376 |
Missense Mutation |
GAG,GAT |
E76D |
NP_001004339.2 |
NM_001307931.1 |
376 |
UTR 5 |
|
|
NP_001294860.1 |
XM_011541480.2 |
376 |
Missense Mutation |
GAG,GAT |
E76D |
XP_011539782.1 |
XM_011541481.2 |
376 |
Missense Mutation |
GAG,GAT |
E76D |
XP_011539783.1 |
XM_011541482.2 |
376 |
Missense Mutation |
GAG,GAT |
E76D |
XP_011539784.1 |
XM_011541483.2 |
376 |
Missense Mutation |
GAG,GAT |
E76D |
XP_011539785.1 |
XM_011541484.1 |
376 |
Missense Mutation |
GAG,GAT |
E76D |
XP_011539786.1 |
XM_011541485.1 |
376 |
Missense Mutation |
GAG,GAT |
E76D |
XP_011539787.1 |
XM_011541486.1 |
376 |
Missense Mutation |
GAG,GAT |
E76D |
XP_011539788.1 |
XM_011541487.2 |
376 |
Missense Mutation |
GAG,GAT |
E76D |
XP_011539789.1 |
XM_011541488.1 |
376 |
Missense Mutation |
GAG,GAT |
E76D |
XP_011539790.1 |
View Full Product Details