Product Details

SNP ID
rs686038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40898791 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGCCTGTATTCAGCATTTTATGC[A/G]TAGATGTATATTTCAGGGAAGTAAG
Phenotype
MIM: 608551
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105370943 PubMed Links

Gene Details

Gene
LOC105370943
Gene Name
uncharacterized LOC105370943
There are no transcripts associated with this gene.

Gene
VPS18
Gene Name
VPS18, CORVET/HOPS core subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020857.2 82 Intron NP_065908.1
XM_011521843.2 82 UTR 5 XP_011520145.1
XM_011521844.2 82 Intron XP_011520146.1
XM_017022447.1 82 Intron XP_016877936.1

View Full Product Details