Product Details
- SNP ID
-
rs750087
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
46
- Location
-
Chr.1:204841033 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGAATTCATTTCCAAAGTGGCAGG[A/G]CTAAGCTATAATGTTAACCCTCTGA
- Phenotype
-
MIM: 609145
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NFASC
PubMed Links
Gene Details
- Gene
- NFASC
- Gene Name
- neurofascin
View Full Product Details