Product Details

SNP ID
rs121909568
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:173904028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCGAACGGCCAGCAATCACAACA[A/G]CGGTACTTGCAGCTGCTTCACTGCC
Phenotype
MIM: 107300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINC1 PubMed Links

Gene Details

Gene
SERPINC1
Gene Name
serpin family C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000488.3 1375 Missense Mutation GCT,GTT A419V NP_000479.1
XM_005245198.3 1375 Missense Mutation GCT,GTT A371V XP_005245255.1

View Full Product Details