Product Details

SNP ID
rs543283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:125154412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCTCCCAAGTTAGGGAGTGAG[A/G]GGATCGCCCGCCGAGGGGGTGGCTC
Phenotype
MIM: 604069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100126584 PubMed Links

Gene Details

Gene
LOC100126584
Gene Name
uncharacterized LOC100126584
There are no transcripts associated with this gene.

Gene
TPD52L1
Gene Name
tumor protein D52-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003395.2 224 UTR 5 NP_001003395.1
NM_001003396.2 224 Intron NP_001003396.1
NM_001003397.2 224 Intron NP_001003397.1
NM_001292026.2 224 UTR 5 NP_001278955.1
NM_001300994.2 224 Intron NP_001287923.1
NM_001318903.1 224 Intron NP_001305832.1
NM_001318907.1 224 Intron NP_001305836.1
NM_003287.3 224 Intron NP_003278.1
XM_005267122.2 224 Intron XP_005267179.1
XM_006715557.3 224 Intron XP_006715620.1
XM_017011237.1 224 Intron XP_016866726.1
XM_017011238.1 224 Intron XP_016866727.1
XM_017011239.1 224 UTR 5 XP_016866728.1
XM_017011240.1 224 Intron XP_016866729.1

View Full Product Details