Product Details

SNP ID
rs454150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42000353 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGATAGGGATGGCTGGACCAGAGC[A/G]ACCTGAGAAAGGTCTGGATTCTGGA
Phenotype
MIM: 182350 MIM: 600283
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP1A3 PubMed Links

Gene Details

Gene
ATP1A3
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
There are no transcripts associated with this gene.

Gene
GRIK5
Gene Name
glutamate ionotropic receptor kainate type subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301030.1 Intron NP_001287959.1
NM_002088.4 Intron NP_002079.3
XM_005258821.3 Intron XP_005258878.1
XM_011526862.2 Intron XP_011525164.1
XM_011526863.2 Intron XP_011525165.1
XM_011526865.1 Intron XP_011525167.1
XM_011526867.1 Intron XP_011525169.1
XM_011526868.1 Intron XP_011525170.1
XM_011526869.2 Intron XP_011525171.1
XM_011526870.2 Intron XP_011525172.1
XM_011526871.2 Intron XP_011525173.1
XM_017026713.1 Intron XP_016882202.1

View Full Product Details