Product Details

SNP ID
rs508152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173000551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCATGATATAGGAAAATAAATA[C/T]TATCTAAGAATCACGAGACCTAGGT
Phenotype
MIM: 603931
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0E1 PubMed Links

Gene Details

Gene
ATP6V0E1
Gene Name
ATPase H+ transporting V0 subunit e1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003945.3 Intron NP_003936.1

View Full Product Details