Product Details

SNP ID
rs308410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:122831438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCACTACTGCCTTTGGGGTAAA[C/T]AGAACCATTGATTTAATAATACACG
Phenotype
MIM: 134920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF2 PubMed Links

Gene Details

Gene
FGF2
Gene Name
fibroblast growth factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002006.4 Intron NP_001997.5

View Full Product Details