Product Details

SNP ID
rs366192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:122828796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAAAAGATGCTTGTAACCTATTA[C/G]ATGGGTTAGTGCTAGTTCTCTATTC
Phenotype
MIM: 134920
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FGF2 PubMed Links

Gene Details

Gene
FGF2
Gene Name
fibroblast growth factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002006.4 Intron NP_001997.5

View Full Product Details