Product Details

SNP ID
rs630526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:79814627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTGACACTGCCTGCGTGGGTCA[A/G]CATCAATCCTGGCGGTGAGATTGTA
Phenotype
MIM: 605696
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNG2 PubMed Links

Gene Details

Gene
KCNG2
Gene Name
potassium voltage-gated channel modifier subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012283.1 Intron NP_036415.1
XM_011525918.2 Intron XP_011524220.1
XM_011525920.2 Intron XP_011524222.1
XM_017025704.1 Intron XP_016881193.1
XM_017025705.1 Intron XP_016881194.1

View Full Product Details