Product Details

SNP ID
rs1078604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56713370 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGAGCAGTGGTTCTCAACTAGGG[G/T]GTAGTTAGGTCTCCCAGGAGATATA
Phenotype
MIM: 601234
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NACA PubMed Links

Gene Details

Gene
NACA
Gene Name
nascent polypeptide-associated complex alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113201.2 Intron NP_001106672.1
NM_001113202.1 Intron NP_001106673.1
NM_001113203.2 Intron NP_001106674.2
NM_001320193.1 Intron NP_001307122.1
NM_001320194.1 Intron NP_001307123.1
NM_005594.5 Intron NP_005585.1
XM_006719412.1 Intron XP_006719475.1
XM_006719413.2 Intron XP_006719476.1
XM_006719414.1 Intron XP_006719477.1
XM_006719418.1 Intron XP_006719481.1
XM_006719420.3 Intron XP_006719483.1
XM_011538389.1 Intron XP_011536691.1
XM_017019337.1 Intron XP_016874826.1

View Full Product Details