Product Details

SNP ID
rs506911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:93413665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCTTCATATGCCACACACAT[C/T]TAGCCTGATTGGCAAATTGGTTTTT
Phenotype
MIM: 603679
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2N PubMed Links

Gene Details

Gene
UBE2N
Gene Name
ubiquitin conjugating enzyme E2 N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003348.3 Intron NP_003339.1
XM_011538719.2 Intron XP_011537021.1
XM_017019918.1 Intron XP_016875407.1

View Full Product Details