Product Details

SNP ID
rs558749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2065796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCTGGAGGTGCCCGGTCCCCG[C/T]TTGGGGATGGGAGTTTTGTAGACTG
Phenotype
MIM: 610647 MIM: 610887
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAT8L PubMed Links

Gene Details

Gene
NAT8L
Gene Name
N-acetyltransferase 8 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178557.3 2578 UTR 3 NP_848652.2
Gene
POLN
Gene Name
DNA polymerase nu
There are no transcripts associated with this gene.

View Full Product Details