Product Details

SNP ID
rs382701
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:2063529 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTTGGGCTGGCTTTCTCTGCTTAAG[C/G]AGCTCTGGAGGGGGGCTCTACCTGC
Phenotype
MIM: 610647 MIM: 610887
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NAT8L PubMed Links

Gene Details

Gene
NAT8L
Gene Name
N-acetyltransferase 8 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178557.3 Intron NP_848652.2
Gene
POLN
Gene Name
DNA polymerase nu
There are no transcripts associated with this gene.

View Full Product Details