Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001161465.1 | 596 | Intron | NP_001154937.1 | ||
NM_023007.2 | 596 | Intron | NP_075383.2 | ||
XM_011544262.2 | 596 | Intron | XP_011542564.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001323930.1 | 596 | Intron | NP_001310859.1 | ||
NM_001323931.1 | 596 | Intron | NP_001310860.1 | ||
NM_001323932.1 | 596 | Intron | NP_001310861.1 | ||
NM_001323933.1 | 596 | Intron | NP_001310862.1 | ||
NM_001323934.1 | 596 | Intron | NP_001310863.1 | ||
NM_001323935.1 | 596 | Intron | NP_001310864.1 | ||
NM_053052.3 | 596 | Intron | NP_444280.2 | ||
XM_006711734.3 | 596 | Intron | XP_006711797.1 | ||
XM_017000231.1 | 596 | UTR 5 | XP_016855720.1 | ||
XM_017000232.1 | 596 | UTR 5 | XP_016855721.1 |