Product Details

SNP ID
rs465770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:98857833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAAATTTCCTAAACCAAACCTG[A/G]ATCACTATTATATCAAGCTTTTATC
Phenotype
MIM: 602118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHD1 PubMed Links

Gene Details

Gene
CHD1
Gene Name
chromodomain helicase DNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270.2 Intron NP_001261.2
XM_005271866.4 Intron XP_005271923.1
XM_005271867.4 Intron XP_005271924.1
XM_011543112.2 Intron XP_011541414.1
XM_017008991.1 Intron XP_016864480.1
XM_017008992.1 Intron XP_016864481.1

View Full Product Details