Product Details

SNP ID
rs275832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:64698359 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAATCGCTTCCTATCTCTTTCAAT[A/G]GAGAAAACAGTCTTCATCATGACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM159B PubMed Links

Gene Details

Gene
FAM159B
Gene Name
family with sequence similarity 159 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164442.1 Intron NP_001157914.1

View Full Product Details